Assay Details
Gene Symbol: NPFFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962137.1 | XP_027817938.1 | 4 - 5 | 641 | 71 | |
GenBank mRNA | EU045356.1 | - | 1 - 2 | 83 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
