Assay Details
Gene Symbol: DOHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008617.4 | XP_004008666.2 | 2 - 3 | 654 | 63 | |
XM_027969887.1 | XP_027825688.1 | 2 - 3 | 624 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
