Assay Details
Gene Symbol: LOC114108792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956275.1 | XP_027812076.1 | 1 | 342 | 82 |
Gene Symbol: ADSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027976332.1 | XP_027832133.1 | 2 - 3 | 392 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
