Assay Details
Gene Symbol: CDPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007051.3 | XP_004007100.1 | 2 - 3 | 203 | 93 | |
XM_012175783.3 | XP_012031173.1 | 2 - 3 | 823 | 93 | ||
XM_012175784.3 | XP_012031174.1 | 2 - 3 | 534 | 93 | ||
XM_012175785.3 | XP_012031175.1 | 2 - 3 | 397 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
