Assay Details
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017912.4 | XP_004017961.2 | 20 - 21 | 1867 | 85 | |
XM_012098847.3 | XP_011954237.2 | 26 - 27 | 3690 | 85 | ||
XM_012098849.3 | XP_011954239.2 | 25 - 26 | 3663 | 85 | ||
XM_012098850.3 | XP_011954240.2 | 25 - 26 | 3561 | 85 | ||
XM_012098851.3 | XP_011954241.2 | 24 - 25 | 3534 | 85 | ||
XM_012098852.3 | XP_011954242.2 | 21 - 22 | 1816 | 85 | ||
XM_012098853.3 | XP_011954243.2 | 21 - 22 | 2884 | 85 | ||
XM_012098854.3 | XP_011954244.2 | 21 - 22 | 1894 | 85 | ||
XM_012098855.3 | XP_011954245.2 | 21 - 22 | 2764 | 85 | ||
XM_012098857.3 | XP_011954247.2 | 21 - 22 | 2755 | 85 | ||
XM_012098858.3 | XP_011954248.2 | 19 - 20 | 1738 | 85 | ||
XM_027957293.1 | XP_027813094.1 | 20 - 21 | 1765 | 85 | ||
XM_027957294.1 | XP_027813095.1 | 20 - 21 | 2754 | 85 | ||
XM_027957295.1 | XP_027813096.1 | 20 - 21 | 2626 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
