Assay Details
Gene Symbol: LOC101106975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957845.1 | XP_027813646.1 | 3 - 4 | 226 | 82 | |
XM_027957846.1 | XP_027813647.1 | 5 - 6 | 341 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
