Assay Details
Gene Symbol: LOC101106975 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957845.1 | XP_027813646.1 | 5 - 6 | 459 | 60 | |
XM_027957846.1 | XP_027813647.1 | 7 - 8 | 574 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
