Assay Details
Gene Symbol: HPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960548.1 | XP_027816349.1 | 6 - 7 | 673 | 61 | |
XM_027960550.1 | XP_027816351.1 | 7 - 8 | 796 | 61 | ||
XM_027960551.1 | XP_027816352.1 | 7 - 8 | 821 | 61 | ||
XM_027960552.1 | XP_027816353.1 | 6 - 7 | 664 | 61 | ||
XM_027960553.1 | XP_027816354.1 | 5 - 6 | 547 | 61 | ||
XM_027960554.1 | XP_027816355.1 | 6 - 7 | 742 | 61 | ||
XM_027960556.1 | XP_027816357.1 | 6 - 7 | 742 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
