Assay Details
Gene Symbol: ECPAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092979.2 | XP_014948465.2 | 31 - 32 | 3581 | 93 | |
XM_015092980.2 | XP_014948466.2 | 30 - 31 | 3504 | 93 | ||
XM_027964118.1 | XP_027819919.1 | 30 - 31 | 3764 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
