Assay Details
Gene Symbol: BMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012188312.3 | XP_012043702.1 | 5 - 6 | 419 | 77 | |
XM_027976604.1 | XP_027832405.1 | 5 - 6 | 549 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
