Assay Details
Gene Symbol: LOC101107224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012097572.2 | XP_011952962.2 | 5 - 6 | 792 | 98 | |
XM_027956332.1 | XP_027812133.1 | 5 - 6 | 792 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
