Assay Details
Gene Symbol: FHL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012182752.3 | XP_012038142.1 | 2 - 3 | 334 | 62 | |
XM_027972435.1 | XP_027828236.1 | 2 - 3 | 320 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
