Assay Details
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021054.3 | XP_004021103.1 | 7 - 8 | 876 | 62 | |
XM_012104638.3 | XP_011960028.1 | 7 - 8 | 879 | 62 | ||
XM_027961728.1 | XP_027817529.1 | 6 - 7 | 1689 | 62 | ||
XM_027961729.1 | XP_027817530.1 | 6 - 7 | 1690 | 62 | ||
XM_027961730.1 | XP_027817531.1 | 6 - 7 | 1686 | 62 | ||
XM_027961731.1 | XP_027817532.1 | 7 - 8 | 876 | 62 | ||
XM_027961732.1 | XP_027817533.1 | 6 - 7 | 1692 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
