Assay Details
Gene Symbol: AKAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022078.4 | XP_004022127.1 | 3 - 4 | 926 | 76 | |
XM_012106705.3 | XP_011962095.1 | 3 - 4 | 1107 | 76 | ||
XM_012106707.3 | XP_011962097.1 | 3 - 4 | 259 | 76 | ||
XM_027963040.1 | XP_027818841.1 | 3 - 4 | 453 | 76 | ||
XM_027963041.1 | XP_027818842.1 | 2 - 3 | 543 | 76 | ||
XM_027963042.1 | XP_027818843.1 | 4 - 5 | 938 | 76 | ||
XM_027963043.1 | XP_027818844.1 | 5 - 6 | 1332 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
