Assay Details
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002014.4 | XP_004002063.1 | 1 - 2 | 724 | 65 | |
XM_012154685.3 | XP_012010075.1 | 1 - 2 | 867 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
