Assay Details
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002014.4 | XP_004002063.1 | 2 - 3 | 882 | 80 | |
XM_012154685.3 | XP_012010075.1 | 2 - 3 | 1025 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
