Assay Details
Gene Symbol: NXPE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016039.4 | XP_004016088.3 | 3 - 4 | 1012 | 108 | |
XM_012095480.3 | XP_011950870.2 | 3 - 4 | 1012 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
