Assay Details
Gene Symbol: C6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017016.4 | XP_004017065.2 | 11 - 12 | 2014 | 54 | |
XM_012096997.3 | XP_011952387.2 | 11 - 12 | 2066 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
