Assay Details
Gene Symbol: WBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020150.3 | XP_004020199.1 | 3 - 4 | 3114 | 92 | |
XM_027960546.1 | XP_027816347.1 | 3 - 4 | 647 | 92 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020150.3 | XP_004020199.1 | 3 - 4 | 3114 | 92 | |
XM_027960546.1 | XP_027816347.1 | 3 - 4 | 647 | 92 |