Assay Details
Gene Symbol: CYRIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012164859.3 | XP_012020249.1 | 5 - 6 | 973 | 87 | |
XM_015094234.2 | XP_014949720.1 | 5 - 6 | 514 | 87 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CYRIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012164859.3 | XP_012020249.1 | 5 - 6 | 973 | 87 | |
XM_015094234.2 | XP_014949720.1 | 5 - 6 | 514 | 87 |