Assay Details
Gene Symbol: SPINDOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959293.1 | XP_027815094.1 | 2 - 3 | 2615 | 85 | |
XM_027959294.1 | XP_027815095.1 | 3 - 4 | 752 | 85 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SPINDOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959293.1 | XP_027815094.1 | 2 - 3 | 2615 | 85 | |
XM_027959294.1 | XP_027815095.1 | 3 - 4 | 752 | 85 |