Assay Details
Gene Symbol: PWWP3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008777.4 | XP_004008826.2 | 9 - 10 | 1407 | 72 | |
XM_012178453.2 | XP_012033843.2 | 9 - 10 | 1429 | 72 | ||
XM_012178454.3 | XP_012033844.2 | 9 - 10 | 1328 | 72 | ||
XM_027970187.1 | XP_027825988.1 | 9 - 10 | 1328 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
