Assay Details
Gene Symbol: SLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020113.4 | XP_004020162.1 | 14 - 15 | 3026 | 65 | |
XM_012102799.3 | XP_011958189.1 | 14 - 15 | 3023 | 65 | ||
XM_027960539.1 | XP_027816340.1 | 10 - 11 | 1547 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
