Assay Details
Gene Symbol: ST14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959398.1 | XP_027815199.1 | 13 - 14 | 4231 | 66 | |
XM_027959399.1 | XP_027815200.1 | 14 - 15 | 4147 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
