Assay Details
Gene Symbol: MOGS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006098.4 | XP_004006147.4 | 3 - 4 | 915 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
