Assay Details
Gene Symbol: TENT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012189745.3 | XP_012045135.3 | 6 - 7 | 1185 | 171 | |
XM_027977819.1 | XP_027833620.1 | 6 - 7 | 1185 | 171 |

More Information
Set Membership: |
![]() ![]() ![]() |
