Assay Details
Gene Symbol: SPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100076.2 | XP_014955562.2 | 5 - 6 | 1159 | 87 | |
XM_027977522.1 | XP_027833323.1 | 2 - 3 | 2726 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
