Assay Details
Gene Symbol: LOC114110160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960067.1 | XP_027815868.1 | 10 - 11 | 2031 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC114110160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960067.1 | XP_027815868.1 | 10 - 11 | 2031 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |