Assay Details
Gene Symbol: ERG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961759.1 | XP_027817560.1 | 9 - 10 | 1448 | 75 | |
XM_027961764.1 | XP_027817565.1 | 9 - 10 | 968 | 75 | ||
XM_027961766.1 | XP_027817567.1 | 9 - 10 | 1073 | 75 | ||
XM_027961773.1 | XP_027817574.1 | 8 - 9 | 1375 | 75 | ||
XM_027961778.1 | XP_027817579.1 | 8 - 9 | 998 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
