Assay Details
Gene Symbol: GDF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012176070.3 | XP_012031460.3 | 2 - 3 | 853 | 93 | |
GenBank mRNA | KC561774.1 | - | 1 - 2 | 659 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
