Assay Details
Gene Symbol: RETREG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012940.4 | XP_004012989.1 | 2 - 3 | 474 | 69 | |
XM_012186125.3 | XP_012041515.1 | 2 - 3 | 484 | 69 | ||
XM_027974489.1 | XP_027830290.1 | 2 - 3 | 479 | 69 | ||
XM_027974490.1 | XP_027830291.1 | 3 - 4 | 320 | 69 | ||
XM_027974491.1 | XP_027830292.1 | 3 - 4 | 225 | 69 | ||
XM_027974492.1 | XP_027830293.1 | 3 - 4 | 166 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
