Assay Details
Gene Symbol: RETREG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012940.4 | XP_004012989.1 | 3 - 4 | 498 | 54 | |
XM_012186125.3 | XP_012041515.1 | 3 - 4 | 508 | 54 | ||
XM_027974489.1 | XP_027830290.1 | 3 - 4 | 503 | 54 | ||
XM_027974490.1 | XP_027830291.1 | 4 - 5 | 344 | 54 | ||
XM_027974491.1 | XP_027830292.1 | 4 - 5 | 249 | 54 | ||
XM_027974492.1 | XP_027830293.1 | 4 - 5 | 190 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
