Assay Details
Gene Symbol: RETREG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012940.4 | XP_004012989.1 | 5 - 6 | 717 | 73 | |
XM_012186125.3 | XP_012041515.1 | 5 - 6 | 727 | 73 | ||
XM_027974489.1 | XP_027830290.1 | 5 - 6 | 722 | 73 | ||
XM_027974490.1 | XP_027830291.1 | 6 - 7 | 563 | 73 | ||
XM_027974491.1 | XP_027830292.1 | 6 - 7 | 468 | 73 | ||
XM_027974492.1 | XP_027830293.1 | 6 - 7 | 409 | 73 | ||
XM_027974493.1 | XP_027830294.1 | 2 - 3 | 732 | 73 | ||
XM_027974494.1 | XP_027830295.1 | 2 - 3 | 472 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027974493.1, XM_027974494.1 transcripts.
Set Membership: |
![]() ![]() |
