Assay Details
Gene Symbol: ASPHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101695.2 | XP_014957181.2 | 2 - 3 | 1145 | 73 | |
XM_027956673.1 | XP_027812474.1 | 2 - 3 | 1145 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
