Assay Details
Gene Symbol: ZRANB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012138942.3 | XP_011994332.2 | 10 - 11 | 1473 | 84 | |
XM_027965011.1 | XP_027820812.1 | 10 - 11 | 1246 | 84 |
Gene Symbol: LOC114112015 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963701.1 | XP_027819502.1 | 2 - 3 | 506 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
