Assay Details
Gene Symbol: ADA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006971.4 | XP_004007020.4 | 3 - 4 | 634 | 63 | |
XM_012175611.3 | XP_012031001.3 | 3 - 4 | 634 | 63 | ||
XM_027968053.1 | XP_027823854.1 | 3 - 4 | 615 | 63 | ||
XM_027968054.1 | XP_027823855.1 | 3 - 4 | 634 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
