Assay Details
Gene Symbol: BICRAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018824.4 | XP_004018873.2 | 5 - 6 | 6205 | 100 | |
XM_012100682.2 | XP_011956072.2 | 5 - 6 | 1872 | 100 | ||
XM_015102717.2 | XP_014958203.2 | 5 - 6 | 1912 | 100 | ||
XM_027958601.1 | XP_027814402.1 | 2 - 3 | 3751 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
