Assay Details
Gene Symbol: BICRAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018824.4 | XP_004018873.2 | 8 - 9 | 6474 | 65 | |
XM_012100682.2 | XP_011956072.2 | 8 - 9 | 2141 | 65 | ||
XM_015102717.2 | XP_014958203.2 | 8 - 9 | 2181 | 65 | ||
XM_027958601.1 | XP_027814402.1 | 5 - 6 | 4020 | 65 | ||
XM_027958602.1 | XP_027814403.1 | 5 - 6 | 7099 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
