Assay Details
Gene Symbol: TAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104843.2 | XP_014960329.1 | 5 - 6 | 643 | 62 | |
XM_015104845.2 | XP_014960331.1 | 5 - 6 | 643 | 62 | ||
XM_015104846.2 | XP_014960332.1 | 5 - 6 | 643 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
