Assay Details
Gene Symbol: DUOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971860.1 | XP_027827661.1 | 2 - 3 | 337 | 73 | |
XM_027971861.1 | XP_027827662.1 | 2 - 3 | 340 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
