Assay Details
Gene Symbol: DUOXA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971860.1 | XP_027827661.1 | 4 - 5 | 526 | 73 | |
XM_027971861.1 | XP_027827662.1 | 4 - 5 | 529 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
