Assay Details
Gene Symbol: PROCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012189121.3 | XP_012044511.2 | 2 - 3 | 492 | 100 | |
XM_015099846.2 | XP_014955332.2 | 3 - 4 | 484 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
