Assay Details
Gene Symbol: LOC101122444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011715.4 | XP_004011764.2 | 12 - 13 | 1095 | 112 | |
XM_012183791.3 | XP_012039181.1 | 12 - 13 | 1081 | 112 | ||
XM_012183794.3 | XP_012039184.1 | 11 - 12 | 1011 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() |
