Assay Details
Gene Symbol: ZNF644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002185.4 | XP_004002234.1 | 3 - 4 | 233 | 68 | |
XM_012175813.3 | XP_012031203.2 | 6 - 7 | 4006 | 68 | ||
XM_015092096.2 | XP_014947582.2 | 6 - 7 | 7564 | 68 | ||
XM_015092097.2 | XP_014947583.2 | 7 - 8 | 7164 | 68 | ||
XM_015092098.2 | XP_014947584.2 | 6 - 7 | 4967 | 68 | ||
XM_015092104.2 | XP_014947590.2 | 5 - 6 | 7439 | 68 | ||
XM_015092105.2 | XP_014947591.1 | 3 - 4 | 3792 | 68 | ||
XM_015092106.2 | XP_014947592.1 | 3 - 4 | 1195 | 68 | ||
XM_027971944.1 | XP_027827745.1 | 6 - 7 | 3949 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
