Assay Details
Gene Symbol: CBARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970174.1 | XP_027825975.1 | 5 - 6 | 512 | 69 | |
XM_027970176.1 | XP_027825977.1 | 3 - 4 | 469 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
