Assay Details
Gene Symbol: TASOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099687.2 | XP_014955173.2 | 22 - 23 | 11872 | 116 | |
XM_015099688.2 | XP_014955174.2 | 22 - 23 | 11853 | 116 | ||
XM_015099691.2 | XP_014955177.2 | 21 - 22 | 11628 | 116 | ||
XM_027976780.1 | XP_027832581.1 | 22 - 23 | 11872 | 116 | ||
XM_027976781.1 | XP_027832582.1 | 23 - 24 | 8302 | 116 | ||
XM_027976782.1 | XP_027832583.1 | 16 - 17 | 6995 | 116 | ||
XM_027976783.1 | XP_027832584.1 | 21 - 22 | 11610 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
