Assay Details
Gene Symbol: ITPRID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004533.4 | XP_004004582.3 | 11 - 12 | 2803 | 82 | |
XM_004004534.4 | XP_004004583.4 | 11 - 12 | 2802 | 82 | ||
XM_027964773.1 | XP_027820574.1 | 11 - 12 | 2800 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
