Assay Details
Gene Symbol: AADACL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013803.3 | XP_004013852.3 | 2 - 3 | 1961 | 78 | |
XM_027975883.1 | XP_027831684.1 | 5 - 6 | 953 | 78 | ||
XM_027975884.1 | XP_027831685.1 | 4 - 5 | 643 | 78 | ||
XM_027975885.1 | XP_027831686.1 | 6 - 7 | 1075 | 78 | ||
XM_027975886.1 | XP_027831687.1 | 3 - 4 | 443 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
