Assay Details
Gene Symbol: PML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017861.4 | XP_004017910.2 | 7 - 8 | 1796 | 64 | |
XM_012098711.3 | XP_011954101.2 | 7 - 8 | 1796 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
