Assay Details
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004465.4 | XP_004004514.2 | 10 - 11 | 1434 | 89 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004465.4 | XP_004004514.2 | 10 - 11 | 1434 | 89 |